Product Details

SNP ID
rs55868044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23988471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAACAATTTGTTATTGAGCACT[A/G]TCCCCTCATGTCCATATTTGTTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL15 PubMed Links

Gene Details

Gene
KLHL15
Gene Name
kelch like family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030624.2 1733 Missense Mutation ACA,ATA T422I NP_085127.2
XM_006724518.1 1733 Missense Mutation ACA,ATA T422I XP_006724581.1

View Full Product Details