Product Details

SNP ID
rs62307974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39460124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCCCCTGATTCAAGTCAGGACC[A/G]GAACAGTTTTAAACTGTGGCACCTT
Phenotype
MIM: 611135 MIM: 607031 MIM: 603686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLB PubMed Links

Gene Details

Gene
KLB
Gene Name
klotho beta
There are no transcripts associated with this gene.

Gene
LIAS
Gene Name
lipoic acid synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278590.1 Intron NP_001265519.1
NM_001278591.1 Intron NP_001265520.1
NM_001278592.1 Intron NP_001265521.1
NM_006859.3 Intron NP_006850.2
NM_194451.2 Intron NP_919433.1
XM_006713990.3 Intron XP_006714053.1
XM_017007665.1 Intron XP_016863154.1
XM_017007666.1 Intron XP_016863155.1
XM_017007667.1 Intron XP_016863156.1
Gene
RPL9
Gene Name
ribosomal protein L9
There are no transcripts associated with this gene.

View Full Product Details