Product Details

SNP ID
rs58436768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153350855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTTGGCCCCAGTCTACTGGCTA[C/T]TTTCATCATATACAAATGTGAAGAA
Phenotype
MIM: 611422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MND1 PubMed Links

Gene Details

Gene
MND1
Gene Name
meiotic nuclear divisions 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253861.1 Intron NP_001240790.1
NM_032117.3 Intron NP_115493.1
XM_005263275.2 Intron XP_005263332.1
XM_017008688.1 Intron XP_016864177.1

View Full Product Details