Product Details

SNP ID
rs61760167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49459074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCTTCCTGCAGGTGCCCGCTT[G/T]CACTTTCTAAACGCTCTGGCAGAAT
Phenotype
MIM: 609413
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 4399 Missense Mutation NP_000115.1
NM_001277058.1 4399 Intron NP_001263987.1
NM_001277059.1 4399 Intron NP_001263988.1

View Full Product Details