Product Details

SNP ID
rs61758222
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:114706653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAAATACGTGATACTGAGTTT[C/T]CCAGTCATGGTAAAGGGTTTGTGCT
Phenotype
MIM: 164790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRAS PubMed Links

Gene Details

Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002524.4 2265 UTR 3 NP_002515.1

View Full Product Details