Product Details

SNP ID
rs56136567
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:116076379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTCACCAAAGAGTAAAGTTAG[C/T]GTTATTTTGGTTATGTGGAATATTA
Phenotype
MIM: 608275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAB21L3 PubMed Links

Gene Details

Gene
MAB21L3
Gene Name
mab-21 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152367.2 Intron NP_689580.2
XM_011540623.2 Intron XP_011538925.1
XM_011540625.2 Intron XP_011538927.1
XM_011540626.2 Intron XP_011538928.1
XM_017000260.1 Intron XP_016855749.1
Gene
SLC22A15
Gene Name
solute carrier family 22 member 15
There are no transcripts associated with this gene.

View Full Product Details