Product Details

SNP ID
rs56822273
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:119794318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTACTTCTTATGGCCAAAGAC[A/C]CAGCTGTTTCATAGGCTGGAGATGC
Phenotype
MIM: 609846
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
REG4 PubMed Links

Gene Details

Gene
REG4
Gene Name
regenerating family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159352.1 1043 UTR 3 NP_001152824.1
NM_001159353.1 1043 Intron NP_001152825.1
NM_032044.3 1043 UTR 3 NP_114433.1

View Full Product Details