Product Details

SNP ID
hCV89755373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98595731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTCTTCTCTTACTACAGAGTA[C/T]AGTTGTGAATTTGGCTCCGCGAAGT
Phenotype
MIM: 600370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A3 PubMed Links

Gene Details

Gene
SLC25A3
Gene Name
solute carrier family 25 member 3
There are no transcripts associated with this gene.

Gene
SNORA53
Gene Name
small nucleolar RNA, H/ACA box 53
There are no transcripts associated with this gene.

View Full Product Details