Product Details

SNP ID
rs56325358
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:6591355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATCTGAGTGGCCGACACAAGC[C/G]TGGAGTACGGCAGCTGCCAACCACA
Phenotype
MIM: 616262 MIM: 165270
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHL21 PubMed Links

Gene Details

Gene
KLHL21
Gene Name
kelch like family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324309.1 3885 UTR 3 NP_001311238.1
NM_014851.3 3885 UTR 3 NP_055666.2
Gene
ZBTB48
Gene Name
zinc finger and BTB domain containing 48
There are no transcripts associated with this gene.

View Full Product Details