Product Details

SNP ID
rs55900921
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:6221560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGGGACAGTGGTGTGCTGTGC[A/G]TATCGTGGGGGTACTCCTAAGCAGC
Phenotype
MIM: 605851 MIM: 616923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICMT PubMed Links

Gene Details

Gene
ICMT
Gene Name
isoprenylcysteine carboxyl methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012405.3 4148 UTR 3 NP_036537.1
XM_011541140.2 4148 UTR 3 XP_011539442.1
Gene
RNF207
Gene Name
ring finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207396.2 4148 Intron NP_997279.2
XM_011541439.2 4148 Intron XP_011539741.1
XM_017001259.1 4148 Intron XP_016856748.1

View Full Product Details