Product Details

SNP ID
rs62157498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112493160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAATGTCGTTATTTCCACCATTT[A/G]TTCACAGACTGGGTTTTGTAGGTGA
Phenotype
MIM: 608291
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTL PubMed Links

Gene Details

Gene
TTL
Gene Name
tubulin tyrosine ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153712.4 Intron NP_714923.1
XM_005263599.3 Intron XP_005263656.1
XM_011510665.2 Intron XP_011508967.1

View Full Product Details