Product Details

SNP ID
rs55968413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130726450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCTGCACAGAAGGTGCTCCAC[A/G]ATGAGGCTGTGGGTGGTCACTGTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR148 PubMed Links

Gene Details

Gene
GPR148
Gene Name
G protein-coupled receptor 148
There are no transcripts associated with this gene.

Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 Intron NP_001264335.1
XM_011511670.2 Intron XP_011509972.1
XM_017004732.1 Intron XP_016860221.1
XM_017004733.1 Intron XP_016860222.1
XM_017004734.1 Intron XP_016860223.1
XM_017004735.1 Intron XP_016860224.1

View Full Product Details