Product Details

SNP ID
rs59746814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135170159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATGCAGGCGGCCTGAGGCCAG[C/T]GCTGTACCAAGTAGAAGACGGTTCC
Phenotype
MIM: 602536 MIM: 615655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB3GAP1 PubMed Links

Gene Details

Gene
RAB3GAP1
Gene Name
RAB3 GTPase activating protein catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172435.1 3383 UTR 3 NP_001165906.1
NM_012233.2 3383 UTR 3 NP_036365.1
XM_011510823.2 3383 Intron XP_011509125.1
XM_011510825.2 3383 UTR 3 XP_011509127.1
Gene
ZRANB3
Gene Name
zinc finger RANBP2-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286568.1 3383 Intron NP_001273497.1
NM_001286569.1 3383 Intron NP_001273498.1
NM_032143.3 3383 Intron NP_115519.2
XM_005263809.1 3383 Intron XP_005263866.1
XM_006712788.1 3383 Intron XP_006712851.1
XM_011511958.2 3383 Intron XP_011510260.1
XM_011511959.2 3383 Intron XP_011510261.2
XM_011511960.1 3383 Intron XP_011510262.1
XM_011511961.2 3383 Intron XP_011510263.2
XM_011511963.1 3383 Intron XP_011510265.1
XM_011511966.2 3383 Intron XP_011510268.2
XM_017005065.1 3383 Intron XP_016860554.1
XM_017005066.1 3383 Intron XP_016860555.1
XM_017005067.1 3383 Intron XP_016860556.1
XM_017005068.1 3383 Intron XP_016860557.1
XM_017005069.1 3383 Intron XP_016860558.1
XM_017005070.1 3383 Intron XP_016860559.1
XM_017005071.1 3383 Intron XP_016860560.1

View Full Product Details