Product Details

SNP ID
rs62170085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135787999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACGGTTTTTGCTCCCTTAACAAC[C/G]CTTAACAACTCTGAAACTTAAAACA
Phenotype
MIM: 603202 MIM: 611216
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LCT PubMed Links

Gene Details

Gene
LCT
Gene Name
lactase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002299.3 6120 UTR 3 NP_002290.2
XM_017004088.1 6120 Intron XP_016859577.1
Gene
UBXN4
Gene Name
UBX domain protein 4
There are no transcripts associated with this gene.

View Full Product Details