Product Details
- SNP ID
-
rs59029020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100470489 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAAGGAGAAACCAGACTCTCACA[A/G]TGAAACATCTTCGTCCCCAGTTCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NMS
PubMed Links
Gene Details
- Gene
- NMS
- Gene Name
- neuromedin S
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011717.1 |
8 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001011717.1 |
View Full Product Details