Product Details
- SNP ID
-
rs62134042
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32278859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTAGCCTCTACAGATTTCCTTGT[A/G]TTATTCTGAGGAAGCGATTGTGCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YIPF4
PubMed Links
Gene Details
- Gene
- YIPF4
- Gene Name
- Yip1 domain family member 4
View Full Product Details