Product Details

SNP ID
rs61756469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47783340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGCGAAGGCGGCCGTGCCGCCG[C/T]TGCCCCCGGGGCCTCTCCTTCCCCA
Phenotype
MIM: 600678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSH6 PubMed Links

Gene Details

Gene
MSH6
Gene Name
mutS homolog 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000179.2 259 Missense Mutation GCT,GTT A36V NP_000170.1
NM_001281492.1 259 Missense Mutation GCT,GTT A36V NP_001268421.1
NM_001281493.1 259 UTR 5 NP_001268422.1
NM_001281494.1 259 Intron NP_001268423.1
XM_011532798.1 259 Intron XP_011531100.1
XM_011532799.1 259 Intron XP_011531101.1

View Full Product Details