Product Details

SNP ID
rs55713459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25735150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGGCTCTTTTCCTGGTTTCG[A/C]TTTTTCTTCCTGTTACAAGATCTCC
Phenotype
MIM: 612991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASXL2 PubMed Links

Gene Details

Gene
ASXL2
Gene Name
additional sex combs like 2, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018263.4 10815 Intron NP_060733.4
XM_006712039.3 10815 UTR 3 XP_006712102.1
XM_006712040.2 10815 UTR 3 XP_006712103.1
XM_011532950.2 10815 UTR 3 XP_011531252.1
XM_011532951.2 10815 UTR 3 XP_011531253.1
XM_017004429.1 10815 UTR 3 XP_016859918.1
XM_017004430.1 10815 UTR 3 XP_016859919.1

View Full Product Details