Product Details

SNP ID
rs61275549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70900676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCCGCGCGCCGGGCGGAGTCT[A/G]GTGGCGGCGGTGGGCGCTGCGGAGA
Phenotype
MIM: 604295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAX2 PubMed Links

Gene Details

Gene
VAX2
Gene Name
ventral anterior homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012476.2 126 Missense Mutation AGT,GGT S19G NP_036608.1
XM_006711982.3 126 Missense Mutation AGT,GGT S19G XP_006712045.1
XM_011532750.2 126 Missense Mutation AGT,GGT S19G XP_011531052.1
XM_011532751.2 126 Missense Mutation AGT,GGT S19G XP_011531053.1

View Full Product Details