Product Details

SNP ID
rs57772551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74137350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCACCAAGGAGAGAAAGCCCA[C/T]CTTTAGCAGGAGAAAAGGAGACCAA
Phenotype
MIM: 613183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOLA3 PubMed Links

Gene Details

Gene
BOLA3
Gene Name
bolA family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035505.1 Intron NP_001030582.1
NM_212552.2 Intron NP_997717.2

View Full Product Details