Product Details

SNP ID
rs55665451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27434468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTAGTAAGTGCTTTGCCTCCC[C/T]AGGTCATTTTTATCACAGAATACAT
Phenotype
MIM: 607386 MIM: 606010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT172 PubMed Links

Gene Details

Gene
IFT172
Gene Name
intraflagellar transport 172
There are no transcripts associated with this gene.

Gene
KRTCAP3
Gene Name
keratinocyte associated protein 3
There are no transcripts associated with this gene.

Gene
NRBP1
Gene Name
nuclear receptor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321357.1 Intron NP_001308286.1
NM_001321358.1 Intron NP_001308287.1
NM_001321359.1 Intron NP_001308288.1
NM_001321361.1 Intron NP_001308290.1
NM_001321362.1 Intron NP_001308291.1
NM_001321363.1 Intron NP_001308292.1
NM_013392.3 Intron NP_037524.1

View Full Product Details