Product Details

SNP ID
rs60600720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88527862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACCAATTTACATTATGGCCAG[A/C]AGGAGACAGTTACTTCTGCTGCTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TEX37 PubMed Links

Gene Details

Gene
TEX37
Gene Name
testis expressed 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152670.2 Intron NP_689883.1
XM_005264188.3 Intron XP_005264245.3
XM_011532691.1 Intron XP_011530993.1
XM_011532692.1 Intron XP_011530994.1

View Full Product Details