Product Details

SNP ID
rs62274613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:89108601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAAGAGAGAATGTTCATATGCA[A/G]CAAATGGCATTTCCTTGATGTTGTG
Phenotype
MIM: 179611
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA3 PubMed Links

Gene Details

Gene
EPHA3
Gene Name
EPH receptor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005233.5 Intron NP_005224.2
NM_182644.2 Intron NP_872585.1
XM_005264715.2 Intron XP_005264772.1
XM_005264716.2 Intron XP_005264773.1

View Full Product Details