Product Details

SNP ID
rs55811133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:20167479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGTGTGTAGACTATATAAAAA[A/G]TCCATAGTTTTACATTCTATTAGAA
Phenotype
MIM: 609168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGO1 PubMed Links

Gene Details

Gene
SGO1
Gene Name
shugoshin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012409.3 Intron NP_001012409.1
NM_001012410.4 Intron NP_001012410.1
NM_001012411.3 Intron NP_001012411.1
NM_001012412.4 Intron NP_001012412.1
NM_001012413.3 Intron NP_001012413.1
NM_001199251.2 Intron NP_001186180.1
NM_001199252.2 Intron NP_001186181.1
NM_001199253.2 Intron NP_001186182.1
NM_001199254.2 Intron NP_001186183.1
NM_001199255.2 Intron NP_001186184.1
NM_001199256.2 Intron NP_001186185.1
NM_001199257.2 Intron NP_001186186.1
NM_138484.4 Intron NP_612493.1
XM_011533373.2 Intron XP_011531675.1
XM_011533375.2 Intron XP_011531677.1
XM_011533376.2 Intron XP_011531678.1
XM_011533377.2 Intron XP_011531679.1
Gene
SGO1-AS1
Gene Name
SGO1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details