Product Details

SNP ID
rs58435324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53170744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCATTTGGACTTCATGGTGCCAG[A/G]CTCTAGTCAGGGATGTGGGGTCCTT
Phenotype
MIM: 176977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCD PubMed Links

Gene Details

Gene
PRKCD
Gene Name
protein kinase C delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316327.1 Intron NP_001303256.1
NM_006254.3 Intron NP_006245.2
NM_212539.1 Intron NP_997704.1
XM_006713257.3 Intron XP_006713320.1
XM_006713259.2 Intron XP_006713322.1
XM_017006855.1 Intron XP_016862344.1
XM_017006856.1 Intron XP_016862345.1

View Full Product Details