Product Details

SNP ID
rs55756006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58193744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTATCCCACACAACAATGACTTG[C/T]GTACTGAGAACGCTGTTCCTTTTTC
Phenotype
MIM: 602244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L3 PubMed Links

Gene Details

Gene
DNASE1L3
Gene Name
deoxyribonuclease I like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256560.1 Intron NP_001243489.1
NM_004944.3 Intron NP_004935.1

View Full Product Details