Product Details

SNP ID
rs56947576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52875355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGGGGGCGTGATGTGCTTTGC[A/G]AGGGCCTGGTTAGGTTTTGCACAAG
Phenotype
MIM: 612404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASL11B PubMed Links

Gene Details

Gene
RASL11B
Gene Name
RAS like family 11 member B
There are no transcripts associated with this gene.

Gene
SCFD2
Gene Name
sec1 family domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152540.3 Intron NP_689753.2
XM_011534375.2 Intron XP_011532677.1
XM_011534376.2 Intron XP_011532678.1
XM_011534378.2 Intron XP_011532680.1
XM_017007786.1 Intron XP_016863275.1
XM_017007787.1 Intron XP_016863276.1

View Full Product Details