Product Details
- SNP ID
-
hCV90066468
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:171390128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGAGCACCAGTTATCTTTAAGA[A/G]CGGTACTTAAACTTTCAAAATAAAC
- Phenotype
-
MIM: 164040
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3912
PubMed Links
Gene Details
- Gene
- MIR3912
- Gene Name
- microRNA 3912
There are no transcripts associated with this gene.
- Gene
- NPM1
- Gene Name
- nucleophosmin
View Full Product Details