Product Details

SNP ID
rs57598427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176528910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGACCCTGGCACCAGCTCTGGA[A/G]ATGCAGGCAGGAGCGAAGGGGCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF44 PubMed Links

Gene Details

Gene
RNF44
Gene Name
ring finger protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014901.4 1833 UTR 3 NP_055716.1
XM_005265840.2 1833 UTR 3 XP_005265897.1
XM_005265841.4 1833 UTR 3 XP_005265898.1
XM_005265842.4 1833 UTR 3 XP_005265899.1
XM_005265843.2 1833 UTR 3 XP_005265900.1
XM_005265844.3 1833 UTR 3 XP_005265901.1
XM_005265845.4 1833 UTR 3 XP_005265902.1
XM_006714831.2 1833 UTR 3 XP_006714894.1
XM_006714832.2 1833 UTR 3 XP_006714895.1
XM_011534466.2 1833 UTR 3 XP_011532768.1
XM_011534467.2 1833 UTR 3 XP_011532769.1

View Full Product Details