Product Details

SNP ID
rs59666856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180300804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAAACTACTCTCTTGCATGGG[A/C]TATTTCAAGCTGTTTTACTATGGGC
Phenotype
MIM: 603865 MIM: 602896
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GFPT2 PubMed Links

Gene Details

Gene
GFPT2
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005110.3 2864 UTR 3 NP_005101.1
XM_006714942.2 2864 UTR 3 XP_006715005.1
XM_017010100.1 2864 UTR 3 XP_016865589.1
Gene
MAPK9
Gene Name
mitogen-activated protein kinase 9
There are no transcripts associated with this gene.

View Full Product Details