Product Details

SNP ID
rs59062625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24914180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGAGCAGCAGAATAAGGAGAT[G/T]GAGGCGGAGATCCAGGCGCTGCGGC
Phenotype
MIM: 162250
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NEFM PubMed Links

Gene Details

Gene
NEFM
Gene Name
neurofilament, medium polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105541.1 420 Intron NP_001099011.1
NM_005382.2 420 Missense Mutation ATG,ATT M129I NP_005373.2

View Full Product Details