Product Details

SNP ID
rs56326305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143598056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCGCGTGGCCGTGAAGATGGC[A/C]TTCCGCAAGGCCTACTCCATCAAGG
Phenotype
MIM: 130592 MIM: 616408
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005250820.3 153 Intron XP_005250877.1
XM_005250823.1 153 Intron XP_005250880.1
XM_006716519.3 153 Intron XP_006716582.1
XM_006716520.3 153 Intron XP_006716583.1
XM_006716522.1 153 Intron XP_006716585.1
XM_006716523.1 153 Intron XP_006716586.1
XM_006716524.1 153 Intron XP_006716587.1
XM_006716525.1 153 Intron XP_006716588.1
XM_011516905.2 153 Intron XP_011515207.1
XM_011516906.2 153 Intron XP_011515208.1
XM_017013168.1 153 Intron XP_016868657.1
XM_017013169.1 153 Intron XP_016868658.1
XM_017013170.1 153 Intron XP_016868659.1
XM_017013171.1 153 Intron XP_016868660.1
XM_017013172.1 153 Intron XP_016868661.1
XM_017013173.1 153 Intron XP_016868662.1
Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TIGD5
Gene Name
tigger transposable element derived 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032862.4 153 Silent Mutation GCA,GCC A51A NP_116251.4

View Full Product Details