Product Details

SNP ID
rs62524242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143716422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCATTGTCCGGAGATACCTACT[C/T]AGGCGGCAGCTCGGGCAGGGGGTGA
Phenotype
MIM: 611927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC166 PubMed Links

Gene Details

Gene
CCDC166
Gene Name
coiled-coil domain containing 166
There are no transcripts associated with this gene.

Gene
FAM83H
Gene Name
family with sequence similarity 83 member H
There are no transcripts associated with this gene.

Gene
LOC101928160
Gene Name
uncharacterized LOC101928160
There are no transcripts associated with this gene.

Gene
MAPK15
Gene Name
mitogen-activated protein kinase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139021.2 86 Silent Mutation CTC,CTT L15L NP_620590.2
XM_011516925.2 86 Intron XP_011515227.1
XM_011516926.2 86 Silent Mutation CTC,CTT L15L XP_011515228.1
XM_011516927.2 86 Intron XP_011515229.1
XM_011516928.2 86 Intron XP_011515230.1
XM_017013218.1 86 Intron XP_016868707.1

View Full Product Details