Product Details

SNP ID
rs57380867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6959814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTGTCATAGACTCAAAAATCTT[C/T]GGAGAAAAGTCATCCCTGGAGTATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFA10P PubMed Links

Gene Details

Gene
DEFA10P
Gene Name
defensin alpha 10, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA8P
Gene Name
defensin alpha 8, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA9P
Gene Name
defensin alpha 9, pseudogene
There are no transcripts associated with this gene.

View Full Product Details