Product Details

SNP ID
rs61755088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83970287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAAGCTCCCGACTCATGACGGAT[C/T]TGTTTAATCCGCTGACCACCTTTGC
Phenotype
MIM: 600712 MIM: 615239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPK PubMed Links

Gene Details

Gene
HNRNPK
Gene Name
heterogeneous nuclear ribonucleoprotein K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318186.1 Intron NP_001305115.1
NM_001318187.1 Intron NP_001305116.1
NM_001318188.1 Intron NP_001305117.1
NM_002140.4 Intron NP_002131.2
NM_031262.3 Intron NP_112552.1
NM_031263.3 Intron NP_112553.1
XM_005251960.2 Intron XP_005252017.1
XM_005251963.3 Intron XP_005252020.1
XM_005251965.2 Intron XP_005252022.1
XM_011518616.1 Intron XP_011516918.1
XM_017014668.1 Intron XP_016870157.1
XM_017014669.1 Intron XP_016870158.1
Gene
MIR7-1
Gene Name
microRNA 7-1
There are no transcripts associated with this gene.

View Full Product Details