Product Details

SNP ID
rs61959940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49701072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGGCTGAAGAGAGTGGTCAAA[C/T]AGGGCTTGCTGTTCTCAAAAATTAG
Phenotype
MIM: 601892
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EBPL PubMed Links

Gene Details

Gene
EBPL
Gene Name
emopamil binding protein like
There are no transcripts associated with this gene.

Gene
KPNA3
Gene Name
karyopherin subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002267.3 2392 UTR 3 NP_002258.2
XM_017020561.1 2392 UTR 3 XP_016876050.1

View Full Product Details