Product Details

SNP ID
rs56132848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72719021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGCTTCTGGGTTAATTCCTTG[C/T]TCACAAAGCCGTACACAAATAGACA
Phenotype
MIM: 610510 MIM: 613448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
There are no transcripts associated with this gene.

Gene
MZT1
Gene Name
mitotic spindle organizing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071775.2 354 Silent Mutation GAA,GAG E52E NP_001065243.1

View Full Product Details