Product Details

SNP ID
rs57857456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:83878768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAAATGGCACTTTCTGATTATAC[C/T]GTATTTTGTTTATAGAAAGTTTGAT
Phenotype
MIM: 609678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLITRK1 PubMed Links

Gene Details

Gene
SLITRK1
Gene Name
SLIT and NTRK like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281503.1 3626 UTR 3 NP_001268432.1
NM_052910.2 3626 UTR 3 NP_443142.1

View Full Product Details