Product Details
- SNP ID
-
hCV90202726
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:111047288 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGTTGTGCTTGTAGGCAATGCA[C/G]CCCAGGATGGCCTTGCTCTGGGTGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GSTM2P1
PubMed Links
Gene Details
- Gene
- GSTM2P1
- Gene Name
- glutathione S-transferase mu 2 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details