Product Details

SNP ID
rs56398248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116791273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGCCCACAGGACAAAGACCCA[C/T]GTTCCTTAGCAAGAAACTCCAATGG
Phenotype
MIM: 613572
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPRC6A PubMed Links

Gene Details

Gene
GPRC6A
Gene Name
G protein-coupled receptor class C group 6 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286354.1 5627 Intron NP_001273283.1
NM_001286355.1 5627 Intron NP_001273284.1
NM_148963.3 5627 Intron NP_683766.2
XM_017010475.1 5627 UTR 3 XP_016865964.1
XM_017010476.1 5627 Intron XP_016865965.1

View Full Product Details