Product Details

SNP ID
rs59094105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122789828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTCAAGGGAAGAGGAAGCTGG[G/T]AGTCCGATGTCCAGCACAGCAAGAG
Phenotype
MIM: 602965 MIM: 610728
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FABP7 PubMed Links

Gene Details

Gene
FABP7
Gene Name
fatty acid binding protein 7
There are no transcripts associated with this gene.

Gene
SMPDL3A
Gene Name
sphingomyelin phosphodiesterase acid like 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286138.1 40 Intron NP_001273067.1
NM_006714.4 40 Intron NP_006705.1
XM_005266797.4 40 UTR 5 XP_005266854.1
XM_005266798.2 40 Intron XP_005266855.1
XM_005266799.2 40 Intron XP_005266856.1
XM_005266801.2 40 Intron XP_005266858.1
XM_005266802.2 40 Intron XP_005266859.1
XM_017010203.1 40 Intron XP_016865692.1
XM_017010204.1 40 Intron XP_016865693.1

View Full Product Details