Product Details

SNP ID
rs61971510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52005361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTTTTTGCACTACAAAATGTT[C/T]ATCTTGGATGCTCAGGAACGTCTAA
Phenotype
MIM: 610858 MIM: 605399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf166 PubMed Links

Gene Details

Gene
C14orf166
Gene Name
chromosome 14 open reading frame 166
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016039.2 4487 Intron NP_057123.1
Gene
NID2
Gene Name
nidogen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007361.3 4487 UTR 3 NP_031387.3
XM_005267405.4 4487 UTR 3 XP_005267462.1
XM_005267406.4 4487 UTR 3 XP_005267463.1
XM_005267407.4 4487 UTR 3 XP_005267464.1

View Full Product Details