Product Details

SNP ID
rs55966267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92063214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAACATATTTTCATTAGTAAA[C/T]TATTTTCATGTTCCAGATCACCATC
Phenotype
MIM: 607047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATXN3 PubMed Links

Gene Details

Gene
ATXN3
Gene Name
ataxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127696.1 1623 UTR 3 NP_001121168.1
NM_001127697.2 1623 UTR 3 NP_001121169.2
NM_001164774.1 1623 UTR 3 NP_001158246.1
NM_001164776.1 1623 UTR 3 NP_001158248.1
NM_001164777.1 1623 UTR 3 NP_001158249.1
NM_001164778.1 1623 UTR 3 NP_001158250.1
NM_001164779.1 1623 UTR 3 NP_001158251.1
NM_001164780.1 1623 UTR 3 NP_001158252.1
NM_001164781.1 1623 UTR 3 NP_001158253.1
NM_001164782.1 1623 UTR 3 NP_001158254.1
NM_004993.5 1623 UTR 3 NP_004984.2
NM_030660.4 1623 UTR 3 NP_109376.1

View Full Product Details