Product Details

SNP ID
rs59914641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100898341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTTTTTGTTGCTGTTACCAAC[A/G]ACACTGCAGTGAGCATTCTTGCAGC
Phenotype
MIM: 613648 MIM: 613649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEG8 PubMed Links

Gene Details

Gene
MEG8
Gene Name
maternally expressed 8 (non-protein coding)
There are no transcripts associated with this gene.

Gene
SNORD112
Gene Name
small nucleolar RNA, C/D box 112
There are no transcripts associated with this gene.

View Full Product Details