Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286277.1 | 4341 | UTR 3 | NP_001273206.1 | ||
NM_001286278.1 | 4341 | Intron | NP_001273207.1 | ||
NM_001286279.1 | 4341 | Intron | NP_001273208.1 | ||
NM_006322.5 | 4341 | UTR 3 | NP_006313.1 | ||
XM_005268293.1 | 4341 | Intron | XP_005268350.1 | ||
XM_011537462.2 | 4341 | UTR 3 | XP_011535764.1 | ||
XM_011537463.2 | 4341 | UTR 3 | XP_011535765.1 | ||
XM_011537464.2 | 4341 | UTR 3 | XP_011535766.1 | ||
XM_017020323.1 | 4341 | UTR 3 | XP_016875812.1 |