Product Details

SNP ID
rs55684031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148982978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATGTTGGTCTCAGGCCCAGCT[C/T]TGACATTGACATTGGCATTGACATT
Phenotype
MIM: 608206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3TC2 PubMed Links

Gene Details

Gene
SH3TC2
Gene Name
SH3 domain and tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024577.3 25752 UTR 3 NP_078853.2

View Full Product Details