Product Details

SNP ID
rs61758779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49994503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCTGGGACACAAAGTCTGCCA[A/G]CATTCCCTGGAAAAAAAAAAGAGCT
Phenotype
MIM: 604980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RACGAP1 PubMed Links

Gene Details

Gene
RACGAP1
Gene Name
Rac GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126103.2 1394 Silent Mutation CTG,TTG L351L NP_001119575.1
NM_001126104.2 1394 Intron NP_001119576.1
NM_001319999.1 1394 Intron NP_001306928.1
NM_001320000.1 1394 Silent Mutation CTG,TTG L351L NP_001306929.1
NM_001320001.1 1394 Intron NP_001306930.1
NM_001320002.1 1394 Intron NP_001306931.1
NM_001320003.1 1394 Intron NP_001306932.1
NM_001320004.1 1394 Intron NP_001306933.1
NM_001320005.1 1394 Intron NP_001306934.1
NM_001320006.1 1394 Intron NP_001306935.1
NM_001320007.1 1394 Intron NP_001306936.1
NM_013277.4 1394 Silent Mutation CTG,TTG L351L NP_037409.2
XM_006719359.1 1394 Intron XP_006719422.1
XM_011538238.1 1394 Intron XP_011536540.1
XM_011538243.1 1394 Silent Mutation CTG,TTG L277L XP_011536545.1
XM_017019220.1 1394 Silent Mutation CTG,TTG L351L XP_016874709.1
XM_017019221.1 1394 Intron XP_016874710.1
XM_017019222.1 1394 Intron XP_016874711.1
XM_017019223.1 1394 Intron XP_016874712.1
XM_017019224.1 1394 Intron XP_016874713.1
XM_017019225.1 1394 Intron XP_016874714.1
XM_017019226.1 1394 Intron XP_016874715.1
XM_017019227.1 1394 Intron XP_016874716.1

View Full Product Details