Product Details

SNP ID
rs57929423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53672594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCACCAAGGAGGGAGTGGAGAC[A/G]AACTTGGAGCAGGCGAACATTTTCA
Phenotype
MIM: 603193
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5G2 PubMed Links

Gene Details

Gene
ATP5G2
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002031.2 336 Silent Mutation TTC,TTT F23F NP_001002031.1
NM_005176.5 336 Silent Mutation NP_005167.2
XM_017019460.1 336 Silent Mutation TTC,TTT F64F XP_016874949.1
XM_017019461.1 336 Silent Mutation TTC,TTT F23F XP_016874950.1
XM_017019462.1 336 Silent Mutation TTC,TTT F7F XP_016874951.1

View Full Product Details