Product Details

SNP ID
rs61108169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54580034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGGCTTTTCTTCCTTCTTGG[C/T]ACCCTGGAAGTTAGTCCTCCCACCT
Phenotype
MIM: 171891 MIM: 613246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE1B PubMed Links

Gene Details

Gene
PDE1B
Gene Name
phosphodiesterase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000924.3 1040 Intron NP_000915.1
NM_001165975.2 1040 Intron NP_001159447.1
NM_001288768.1 1040 Intron NP_001275697.1
NM_001288769.1 1040 Intron NP_001275698.1
NM_001315534.1 1040 Intron NP_001302463.1
NM_001315535.1 1040 Intron NP_001302464.1
XM_011538456.1 1040 Intron XP_011536758.1
XM_017019432.1 1040 Intron XP_016874921.1
XM_017019433.1 1040 Intron XP_016874922.1
Gene
PPP1R1A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006741.3 1040 UTR 3 NP_006732.3
XM_005268995.4 1040 Intron XP_005269052.1
XM_006719471.3 1040 Intron XP_006719534.1

View Full Product Details