Product Details

SNP ID
rs58920821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55469753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTACTCATGATGGACATATAA[C/T]GCAAAGGTTTGCAGATGGCAACATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C70 PubMed Links

Gene Details

Gene
OR6C70
Gene Name
olfactory receptor family 6 subfamily C member 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005499.1 386 Missense Mutation CAT,CGT H129R NP_001005499.1

View Full Product Details